siRNA Details
Gene Symbol: NAT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003960.3 | NP_003951.3 | 2 | 679 | ||
GenBank mRNA | AB013094.1 | - | 2 | 680 | ||
AB019551.1 | - | 2 | 638 | |||
AF187813.1 | - | 2 | 689 | |||
BC012626.1 | - | 2 | 667 | 4045439 | ||
CR407610.1 | - | 1 | 512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
