siRNA Details
Gene Symbol: UGT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019093.2 | NP_061966.1 | 1 | 200 | ||
GenBank mRNA | AY435138.1 | - | 1 | 200 |
Gene Symbol: UGT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007120.2 | NP_009051.1 | 1 | 229 | ||
GenBank mRNA | AK313623.1 | - | 1 | 243 | ||
AY435139.1 | - | 1 | 200 | |||
BC121036.1 | - | 1 | 200 | 40117585 | ||
BC131623.1 | - | 1 | 193 | |||
BC139784.1 | - | 1 | 220 | 40117089 | ||
DQ364249.1 | - | 1 | 209 | |||
M57951.1 | - | 1 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
