siRNA Details
Gene Symbol: GSTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319059.1 | NP_001305988.1 | 2 | 239 | ||
NM_145740.4 | NP_665683.1 | 2 | 239 | |||
XM_005249034.3 | XP_005249091.1 | 2 | 98 | |||
GenBank mRNA | AK293371.1 | - | 2 | 88 | ||
AK310324.1 | - | 2 | 239 | |||
AL096729.1 | - | 879 | ||||
AY532928.1 | - | 1 | 26 | |||
BC053578.1 | - | 2 | 74 | 5167338 | ||
BC110891.1 | - | 2 | 76 | 5171697 | ||
BG430521.1 | - | 2 | 150 | |||
BI762344.1 | - | 2 | 115 | |||
CR407656.1 | - | 1 | 26 | |||
M14777.1 | - | 2 | 92 | |||
M15872.1 | - | 2 | 81 | |||
M21758.1 | - | 2 | 92 | |||
M25627.1 | - | 2 | 92 | |||
S49975.1 | - | 1 | 67 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
