siRNA Details
Gene Symbol: AKR1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135241.2 | NP_001128713.1 | 1 | 174 | ||
NM_001321027.1 | NP_001307956.1 | 1 | 174 | |||
NM_001354.5 | NP_001345.1 | 3 | 408 | |||
NM_205845.2 | NP_995317.1 | 2 | 308 | |||
GenBank mRNA | AB209017.1 | - | 1 | 62 | ||
AK091194.1 | - | 3 | 390 | |||
AK290304.1 | - | 1 | 51 | |||
AK296419.1 | - | 1 | 23 | |||
AK296686.1 | - | 1 | 21 | |||
AK308385.1 | - | 1 | 22 | |||
BC007024.1 | - | 1 | 17 | 3681970 | ||
BC063574.1 | - | 2 | 275 | 3877666 | ||
DA438648.1 | - | 2 | 280 | |||
DA458030.1 | - | 3 | 303 | |||
U05598.1 | - | 1 | 18 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
