siRNA Details
Gene Symbol: COQ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182476.2 | NP_872282.1 | 3 | 429 | ||
NM_182480.2 | NP_872286.2 | 3 | 432 | |||
XM_006720156.1 | XP_006720219.1 | 3 | 473 | |||
XM_011536807.1 | XP_011535109.1 | 3 | 401 | |||
XM_011536810.2 | XP_011535112.1 | 3 | 401 | |||
XM_017021351.1 | XP_016876840.1 | 3 | 250 | |||
GenBank mRNA | AF132944.1 | - | 3 | 355 | ||
AK130167.1 | - | 3 | 386 | |||
AK222965.1 | - | 3 | 371 | |||
AK295517.1 | - | 3 | 371 | |||
AK296040.1 | - | 3 | 440 | |||
AK296386.1 | - | 3 | 371 | |||
AK307306.1 | - | 3 | 473 | |||
AL521052.3 | - | 3 | 460 | |||
AL557147.3 | - | 3 | 421 | |||
BC014181.2 | - | 3 | 378 | 4621678 | ||
BC014483.1 | - | 3 | 366 | 4866198 | ||
BX342249.2 | - | 3 | 430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
