siRNA Details
Gene Symbol: KLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030047.1 | NP_001025218.1 | 4 | 595 | ||
NM_001030048.1 | NP_001025219.1 | 4 | 466 | |||
NM_001648.2 | NP_001639.1 | 4 | 595 | |||
GenBank mRNA | AF335477.1 | - | 4 | 1018 | ||
AF335478.1 | - | 3 | 1833 | |||
AJ310938.1 | - | 3 | 734 | |||
AJ459782.1 | - | 5 | 452 | |||
AJ512346.1 | - | 4 | 575 | |||
AK312933.1 | - | 4 | 589 | |||
BC005307.1 | - | 4 | 595 | 3950475 | ||
BC050595.2 | - | 4 | 618 | 6199928 | ||
BC056665.1 | - | 4 | 600 | 4296876 | ||
BQ932072.1 | - | 4 | 432 | |||
BT019862.1 | - | 4 | 554 | |||
DQ641251.1 | - | 2 | 986 | |||
M21895.1 | - | 4 | 543 | |||
M21896.1 | - | 4 | 543 | |||
M26663.1 | - | 4 | 578 | |||
U17040.1 | - | 4 | 595 | |||
X05332.1 | - | 4 | 597 | |||
X07730.1 | - | 3 | 859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
