siRNA Details
Gene Symbol: PTPRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002838.4 | NP_002829.3 | 5 | 592 | ||
XM_006711472.3 | XP_006711535.1 | 5 | 553 | |||
XM_006711473.3 | XP_006711536.1 | 4 | 337 | |||
XM_006711474.3 | XP_006711537.1 | 4 | 337 | |||
GenBank mRNA | AK299986.1 | - | 4 | 267 | ||
AK303539.1 | - | 5 | 465 | |||
AK307635.1 | - | 5 | 470 | |||
AK308217.1 | - | 4 | 343 | |||
AK310898.1 | - | 4 | 290 | |||
AY429565.1 | - | 3 | 155 | |||
AY429566.1 | - | 3 | 155 | |||
AY567999.1 | - | 5 | 516 | |||
BC014239.2 | - | 5 | 456 | |||
Y00638.1 | - | 5 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
