siRNA Details
Gene Symbol: PTPRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002838.4 | NP_002829.3 | 32, 33 | 3869 | ||
NM_080921.3 | NP_563578.2 | 29, 30 | 3386 | |||
XM_006711472.3 | XP_006711535.1 | 31, 32 | 3686 | |||
XM_006711473.3 | XP_006711536.1 | 31, 32 | 3614 | |||
XM_006711474.3 | XP_006711537.1 | 30, 31 | 3470 | |||
GenBank mRNA | AK292131.1 | - | 29, 30 | 3259 | ||
Y00062.1 | - | 30, 31 | 3313 | |||
Y00638.1 | - | 32, 33 | 3736 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
