siRNA Details
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320893.1 | NP_001307822.1 | 6, 7 | 1008 | ||
NM_022552.4 | NP_072046.2 | 11, 12 | 1694 | |||
NM_153759.3 | NP_715640.2 | 7, 8 | 978 | |||
NM_175629.2 | NP_783328.1 | 11, 12 | 1765 | |||
NR_135490.1 | 11, 12 | 1765 | ||||
XM_005264175.4 | XP_005264232.1 | 11, 12 | 1628 | |||
XM_005264177.4 | XP_005264234.1 | 6, 7 | 920 | |||
XM_011532662.2 | XP_011530964.1 | 9, 10 | 1630 | |||
XM_011532663.2 | XP_011530965.1 | 12, 13 | 1807 | |||
XM_011532664.2 | XP_011530966.1 | 11, 12 | 1705 | |||
XM_011532666.2 | XP_011530968.1 | 7, 8 | 993 | |||
XM_011532667.2 | XP_011530969.1 | 6, 7 | 810 | |||
XM_017003526.1 | XP_016859015.1 | 11, 12 | 1736 | |||
XM_017003527.1 | XP_016859016.1 | 6, 7 | 964 | |||
GenBank mRNA | AB208833.1 | - | 11, 12 | 1664 | ||
AF067972.2 | - | 11, 12 | 1656 | |||
AF331856.1 | - | 11, 12 | 1644 | |||
AF480163.1 | - | 7, 8 | 974 | |||
AK025230.1 | - | 1, 2 | 18 | |||
BC043617.1 | - | 11, 12 | 1664 | 6150112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
