siRNA Details
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320893.1 | NP_001307822.1 | 17, 18 | 2186 | ||
NM_022552.4 | NP_072046.2 | 22, 23 | 2872 | |||
NM_153759.3 | NP_715640.2 | 18, 19 | 2156 | |||
NM_175629.2 | NP_783328.1 | 22, 23 | 2943 | |||
NR_135490.1 | 23, 24 | 3142 | ||||
XM_005264175.4 | XP_005264232.1 | 22, 23 | 2806 | |||
XM_005264177.4 | XP_005264234.1 | 17, 18 | 2098 | |||
XM_011532662.2 | XP_011530964.1 | 20, 21 | 2808 | |||
XM_011532663.2 | XP_011530965.1 | 23, 24 | 2985 | |||
XM_011532666.2 | XP_011530968.1 | 18, 19 | 2171 | |||
XM_011532667.2 | XP_011530969.1 | 17, 18 | 1988 | |||
XM_017003526.1 | XP_016859015.1 | 22, 23 | 2914 | |||
XM_017003527.1 | XP_016859016.1 | 17, 18 | 2142 | |||
GenBank mRNA | AB208833.1 | - | 23, 24 | 3041 | ||
AF067972.2 | - | 22, 23 | 2834 | |||
AF331856.1 | - | 22, 23 | 2822 | |||
AF480163.1 | - | 18, 19 | 2152 | |||
AK025230.1 | - | 12, 13 | 1196 | |||
BC018214.1 | - | 7, 8 | 725 | |||
BC023612.2 | - | 8, 9 | 924 | |||
BC043617.1 | - | 22, 23 | 2842 | 6150112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
