siRNA Details
Gene Symbol: PRR5-ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181334.5 | NP_851851.3 | 13 | 1431 | ||
GenBank mRNA | AF195968.1 | - | 12 | 1022 | ||
AL355192.1 | - | 15 | 1391 | |||
BC132755.1 | - | 11 | 922 | 40146777 | ||
BC132757.1 | - | 11 | 922 | 40146779 | ||
BC144382.1 | - | 11 | 922 | 9052903 |
Gene Symbol: ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017526.1 | NP_001017526.1 | 11 | 995 | ||
NM_001198726.1 | NP_001185655.1 | 10 | 902 | |||
NM_181335.2 | NP_851852.2 | 10 | 902 | |||
GenBank mRNA | AF544240.1 | - | 9 | 761 | ||
AK001306.1 | - | 11 | 995 | |||
AK022305.1 | - | 9 | 844 | |||
BC010490.2 | - | 8 | 746 | |||
BC048280.1 | - | 10 | 854 | 5190702 | ||
BC059382.1 | - | 10 | 886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
