siRNA Details
Gene Symbol: PPFIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304331.1 | NP_001291260.1 | 8 | 1443 | ||
NM_001304332.1 | NP_001291261.1 | 8 | 1443 | |||
XM_006711586.2 | XP_006711649.1 | 8 | 1473 | |||
XM_017002606.1 | XP_016858095.1 | 8 | 1473 | |||
XM_017002607.1 | XP_016858096.1 | 8 | 1473 | |||
XM_017002608.1 | XP_016858097.1 | 8 | 1473 | |||
XM_017002609.1 | XP_016858098.1 | 8 | 1473 | |||
XM_017002610.1 | XP_016858099.1 | 8 | 1473 | |||
GenBank mRNA | AK295757.1 | - | 2 | 171 | ||
EF428334.1 | - | 12 | 1376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
