siRNA Details
Gene Symbol: GPR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195381.1 | NP_001182310.1 | 4 | 757 | ||
NM_001195382.1 | NP_001182311.1 | 4 | 643 | |||
GenBank mRNA | AK172786.1 | - | 4 | 734 | ||
AK290256.1 | - | 4 | 606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
