siRNA Details
Gene Symbol: OR4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005224.1 | NP_001005224.1 | 1 | 565 | ||
XM_017009338.1 | XP_016864827.1 | 2 | 4782 | |||
GenBank mRNA | BC137547.1 | - | 1 | 584 | 9021547 | |
BC137568.1 | - | 1 | 584 | 9021568 |
Gene Symbol: OR4F16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005277.1 | NP_001005277.1 | 1 | 565 | ||
XM_017002408.1 | XP_016857897.1 | 3 | 3297 | |||
XM_017002409.1 | XP_016857898.1 | 4 | 3215 | |||
XM_017002410.1 | XP_016857899.1 | 2 | 3079 | |||
XM_017002411.1 | XP_016857900.1 | 2 | 3068 |
Gene Symbol: OR4F21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005504.1 | NP_001005504.1 | 1 | 565 |
Gene Symbol: OR4F29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005221.2 | NP_001005221.2 | 1 | 565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
