siRNA Details
Gene Symbol: CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145134.1 | NP_001138606.1 | 13, 14 | 1769 | ||
NM_001145135.1 | NP_001138607.1 | 14, 15 | 1871 | |||
NM_001145137.1 | NP_001138609.1 | 13, 14 | 1895 | |||
NM_004377.3 | NP_004368.1 | 14, 15 | 1832 | |||
NM_152245.2 | NP_689451.1 | 14, 15 | 1871 | |||
NM_152246.2 | NP_689452.1 | 14, 15 | 1832 | |||
GenBank mRNA | AK223174.1 | - | 14, 15 | 1771 | ||
AK291316.1 | - | 14, 15 | 1796 | |||
AK297866.1 | - | 13, 14 | 1681 | |||
AK299395.1 | - | 14, 15 | 1565 | |||
BC142954.1 | - | 13, 14 | 1838 | 8860341 | ||
D87812.1 | - | 13, 14 | 1752 | |||
U62733.1 | - | 13, 14 | 1779 | |||
U66828.1 | - | 14, 15 | 1771 | |||
Y08682.1 | - | 14, 15 | 1784 | |||
Y08683.1 | - | 14, 15 | 1784 |
Gene Symbol: CHKB-CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC131570.1 | - | 16, 17 | 2078 | 40108355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
