siRNA Details
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF206665.1 | - | 3 | 369 | ||
AF206666.1 | - | 2 | 287 | |||
BC051852.1 | - | 3 | 364 | |||
M30038.1 | - | 3 | 376 |
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028059.1 | - | 3 | 371 | 5181815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
