siRNA Details
Gene Symbol: CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145134.1 | NP_001138606.1 | 2 | 156 | ||
NM_001145135.1 | NP_001138607.1 | 2 | 156 | |||
NM_001145137.1 | NP_001138609.1 | 1 | 180 | |||
NM_004377.3 | NP_004368.1 | 2 | 117 | |||
NM_152245.2 | NP_689451.1 | 2 | 156 | |||
NM_152246.2 | NP_689452.1 | 2 | 117 | |||
GenBank mRNA | AK223174.1 | - | 2 | 56 | ||
AK291316.1 | - | 2 | 81 | |||
AK297866.1 | - | 2 | 68 | |||
AK299395.1 | - | 2 | 93 | |||
BC142954.1 | - | 1 | 123 | 8860341 | ||
BG426428.1 | - | 102 | ||||
BI560870.1 | - | 1 | 185 | |||
CB961274.1 | - | 2 | 144 | |||
D87812.1 | - | 1 | 37 | |||
DC373481.1 | - | 2 | 156 | |||
U62733.1 | - | 1 | 64 | |||
U66828.1 | - | 2 | 56 | |||
Y08682.1 | - | 2 | 69 | |||
Y08683.1 | - | 2 | 69 |
Gene Symbol: CHKB-CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027928.2 | 12 | 1588 | |||
GenBank mRNA | AB051457.1 | - | 9 | 2226 | ||
AK301477.1 | - | 11 | 1155 | |||
BC131570.1 | - | 4 | 413 | 40108355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
