siRNA Details
Gene Symbol: MMP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002946.1 | 2, 3 | 299 | |||
GenBank mRNA | AF057062.1 | - | 1, 2 | 150 | ||
AF086353.1 | - | 1, 2 | 45 |
Gene Symbol: MMP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006983.1 | NP_008914.1 | 4, 5 | 630 | ||
XM_017002615.1 | XP_016858104.1 | 3, 4 | 1538 | |||
XM_017002616.1 | XP_016858105.1 | 3, 4 | 1538 | |||
XM_017002617.1 | XP_016858106.1 | 3, 4 | 1538 | |||
GenBank mRNA | AB010961.1 | - | 4, 5 | 619 | ||
AB010962.1 | - | 3, 4 | 711 | |||
AF056200.1 | - | 4, 5 | 607 | |||
AJ005256.1 | - | 4, 5 | 630 | |||
BC025719.1 | - | 4, 5 | 621 | 5228894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
