siRNA Details
Gene Symbol: CDY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004680.2 | NP_004671.1 | 1 | 349 | ||
NM_170723.1 | NP_733841.1 | 1 | 349 | |||
XM_011531512.1 | XP_011529814.1 | 1 | 351 | |||
GenBank mRNA | AF000981.1 | - | 1 | 304 | ||
AF080597.1 | - | 1 | 304 | |||
AK292375.1 | - | 1 | 371 | |||
BC033041.1 | - | 1 | 354 | |||
BC132955.1 | - | 1 | 118 | 40146977 |
Gene Symbol: CDY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003894.1 | NP_001003894.1 | 1 | 349 | ||
NM_001003895.1 | NP_001003895.1 | 1 | 349 | |||
XM_011531484.1 | XP_011529786.1 | 1 | 351 | |||
GenBank mRNA | AK292318.1 | - | 1 | 322 | ||
BC132929.1 | - | 1 | 118 | 40146951 |
Gene Symbol: CDY2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004825.2 | NP_004816.1 | 1 | 351 | ||
GenBank mRNA | AF080598.1 | - | 1 | 351 | ||
AK292233.1 | - | 1 | 324 |
Gene Symbol: CDY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001722.1 | NP_001001722.1 | 1 | 350 | ||
GenBank mRNA | BC069087.1 | - | 1 | 350 | 7216901 | |
BC130426.1 | - | 1 | 119 | 40146457 | ||
BC130428.1 | - | 1 | 119 | 40146459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
