siRNA Details
Gene Symbol: CELA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005747.4 | NP_005738.4 | 1 | 47 | ||
GenBank mRNA | AK225620.1 | - | 1 | 43 | ||
AK308510.1 | - | 1 | 40 | |||
AK308514.1 | - | 1 | 43 | |||
AK308518.1 | - | 1 | 47 | |||
BC005918.1 | - | 1 | 47 | 4295129 | ||
BC007028.1 | - | 1 | 48 | 3950453 | ||
BC008383.1 | - | 1 | 43 | 3949901 | ||
BC015103.1 | - | 1 | 51 | 3950076 | ||
D00306.1 | - | 1 | 49 |
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007352.3 | NP_031378.1 | 1 | 174 | ||
GenBank mRNA | AK092382.1 | - | 1 | 47 | ||
AK315798.1 | - | 1 | 43 | |||
BC005216.1 | - | 1 | 48 | 3949903 | ||
BX499439.1 | - | 1 | 143 | |||
M16630.1 | - | 1 | 19 | |||
M18692.1 | - | 1 | 46 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
