siRNA Details
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 1, 2 | 39 | ||
GenBank mRNA | AF022047.1 | - | 1, 2 | 32 | ||
AM851002.1 | - | 1, 2 | 64 | |||
AY789063.2 | - | 12 | ||||
L76670.1 | - | 1, 2 | 39 | |||
X97231.1 | - | 1, 2 | 87 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
