siRNA Details
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 8 | 919 | ||
GenBank mRNA | AF022047.1 | - | 8 | 912 | ||
AM851002.1 | - | 8 | 944 | |||
L76670.1 | - | 8 | 919 | |||
X97231.1 | - | 8 | 967 |
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 8 | 921 | ||
GenBank mRNA | AF022046.1 | - | 8 | 921 | ||
AF285437.1 | - | 7 | 868 | |||
AM999888.1 | - | 8 | 916 | |||
X89892.1 | - | 8 | 928 | |||
X98858.1 | - | 8 | 908 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291696.1 | NP_001278625.1 | 7 | 643 | ||
NM_001291700.1 | NP_001278629.1 | 6 | 349 | |||
NM_001291701.1 | NP_001278630.1 | 7 | 892 | |||
NM_012312.4 | NP_036444.1 | 8 | 943 | |||
GenBank mRNA | AF074087.1 | - | 6 | 305 | ||
AF285438.1 | - | 7 | 868 | |||
AJ002102.1 | - | 8 | 905 | |||
AJ002103.1 | - | 8 | 908 | |||
AJ002104.1 | - | 7 | 857 | |||
AJ002105.1 | - | 7 | 857 | |||
AM409094.1 | - | 8 | 909 | |||
AY366243.1 | - | 8 | 918 | |||
BC119631.1 | - | 8 | 927 | 40115568 | ||
BC119632.1 | - | 8 | 927 | 40115570 | ||
BC126856.1 | - | 8 | 910 | 40017301 | ||
FR750994.1 | - | 8 | 943 | |||
L41347.1 | - | 8 | 927 | |||
L76667.1 | - | 8 | 625 | |||
L76668.1 | - | 7 | 619 | |||
U24079.1 | - | 8 | 906 |
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 8 | 975 | ||
NM_001281972.1 | NP_001268901.1 | 7 | 924 | |||
NM_012314.5 | NP_036446.3 | 9 | 997 | |||
GenBank mRNA | AF002255.1 | - | 9 | 954 | ||
AJ417554.1 | - | 7 | 866 | |||
AJ417555.1 | - | 7 | 888 | |||
AY366245.1 | - | 8 | 942 | |||
AY388472.1 | - | 8 | 946 | |||
BC096694.1 | - | 8 | 919 | 40026979 | ||
BC101977.1 | - | 8 | 919 | 40004312 | ||
BC103693.1 | - | 8 | 919 | 40004314 | ||
BC108918.1 | - | 9 | 910 | 40017300 | ||
BC127084.1 | - | 8 | 925 | 40120468 | ||
BC144566.1 | - | 8 | 997 | 9053094 | ||
BC171788.1 | - | 8 | 997 | 9054442 | ||
L76671.1 | - | 8 | 919 | |||
U24077.1 | - | 8 | 899 | |||
X94609.1 | - | 8 | 963 |
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 8 | 942 | ||
GenBank mRNA | AF208054.1 | - | 8 | 942 | ||
AF272389.1 | - | 9 | 892 | |||
AY366247.1 | - | 8 | 966 | |||
AY366248.1 | - | 10 | 966 | |||
L76672.1 | - | 8 | 919 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 10 | 1229 | ||
NM_001282170.1 | NP_001269099.1 | 9 | 944 | |||
NM_001282171.1 | NP_001269100.1 | 8 | 908 | |||
GenBank mRNA | AF022044.1 | - | 10 | 1205 | ||
AK301140.1 | - | 10 | 1229 | |||
BC098107.1 | - | 8 | 882 | 40026977 | ||
BC098288.1 | - | 10 | 1203 | 40026980 | ||
BC105679.1 | - | 10 | 1203 | 40004313 | ||
EF370406.1 | - | 9 | 1152 | |||
U73396.1 | - | 10 | 1192 | |||
X97232.1 | - | 9 | 907 | |||
X97233.1 | - | 10 | 1192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
