siRNA Details
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 2, 3 | 170 | ||
NM_001281972.1 | NP_001268901.1 | 2, 3 | 170 | |||
NM_012314.5 | NP_036446.3 | 2, 3 | 170 | |||
GenBank mRNA | AF002255.1 | - | 2, 3 | 127 | ||
AF135563.1 | - | 1 | 18 | |||
AJ417554.1 | - | 1, 2 | 61 | |||
AJ417555.1 | - | 1, 2 | 61 | |||
AJ620615.2 | - | 1, 2 | 54 | |||
AJ620616.2 | - | 1, 2 | 54 | |||
AY102623.1 | - | 2, 3 | 81 | |||
AY366245.1 | - | 2, 3 | 115 | |||
AY366246.1 | - | 2, 3 | 88 | |||
AY388472.1 | - | 2, 3 | 119 | |||
AY789064.2 | - | 2, 3 | 65 | |||
BC096694.1 | - | 2, 3 | 92 | 40026979 | ||
BC101977.1 | - | 2, 3 | 92 | 40004312 | ||
BC103693.1 | - | 2, 3 | 92 | 40004314 | ||
BC108918.1 | - | 2, 3 | 83 | 40017300 | ||
BC125147.1 | - | 1, 2 | 65 | |||
BC127084.1 | - | 2, 3 | 98 | 40120468 | ||
BC144566.1 | - | 2, 3 | 170 | 9053094 | ||
BC171788.1 | - | 2, 3 | 170 | 9054442 | ||
L76671.1 | - | 2, 3 | 92 | |||
U24077.1 | - | 2, 3 | 72 | |||
X94609.1 | - | 2, 3 | 136 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF135560.1 | - | 1 | 18 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
