siRNA Details
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 3 | 309 | ||
XM_011526939.2 | XP_011525241.1 | 3 | 914 | |||
XM_017026782.1 | XP_016882271.1 | 3 | 287 | |||
XM_017026783.1 | XP_016882272.1 | 3 | 914 | |||
GenBank mRNA | AF022045.1 | - | 3 | 278 | ||
AF135560.1 | - | 1 | 208 | |||
AF285431.1 | - | 3 | 304 | |||
AF285432.1 | - | 2 | 231 | |||
AY366237.1 | - | 3 | 307 | |||
AY366238.1 | - | 3 | 253 | |||
AY366239.1 | - | 3 | 301 | |||
AY366240.1 | - | 3 | 309 | |||
AY366241.1 | - | 3 | 308 | |||
AY388471.1 | - | 3 | 301 | |||
AY789055.1 | - | 3 | 304 | |||
AY789056.1 | - | 3 | 271 | |||
BC069344.1 | - | 3 | 304 | 7262296 | ||
KJ699235.1 | - | 3 | 544 | |||
L41267.1 | - | 3 | 303 | |||
U24076.1 | - | 3 | 304 | |||
U24078.1 | - | 3 | 311 | |||
U96189.1 | - | 1 | 208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
