siRNA Details
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 8 | 1533 | ||
GenBank mRNA | BC032422.1 | - | 8 | 1509 | 5220339 | |
BC050730.1 | - | 8 | 1527 | 5219336 | ||
L41268.1 | - | 8 | 1533 | |||
U24074.1 | - | 8 | 1517 | |||
U73395.1 | - | 8 | 1517 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 8 | 1531 | ||
XM_017026782.1 | XP_016882271.1 | 8 | 1509 | |||
GenBank mRNA | L41267.1 | - | 9 | 1525 | ||
U24076.1 | - | 9 | 1526 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 8 | 1522 | ||
GenBank mRNA | U24075.1 | - | 8 | 1507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
