siRNA Details
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 1, 2 | 62 | ||
GenBank mRNA | AF208054.1 | - | 1, 2 | 62 | ||
AF272389.1 | - | 1, 2 | 12 | |||
AY366247.1 | - | 1, 2 | 86 | |||
AY366248.1 | - | 1, 2 | 86 | |||
L76672.1 | - | 1, 2 | 39 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
