siRNA Details
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 8 | 1191 | ||
GenBank mRNA | AF208054.1 | - | 8 | 1191 | ||
AY366247.1 | - | 8 | 1215 | |||
L76672.1 | - | 8 | 1168 |
Gene Symbol: KIR2DL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020535.3 | NP_065396.1 | 8 | 1193 | ||
GenBank mRNA | AF204903.1 | - | 8 | 1193 | ||
AF217487.1 | - | 10 | 1162 |
Gene Symbol: KIR2DL5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018081.1 | NP_001018091.1 | 9 | 1138 | ||
GenBank mRNA | AF271607.1 | - | 9 | 1138 | ||
AF271608.1 | - | 9 | 1138 | |||
AJ544587.1 | - | 9 | 1139 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 8 | 1126 | ||
NM_001291696.1 | NP_001278625.1 | 7 | 892 | |||
NM_001291700.1 | NP_001278629.1 | 6 | 598 | |||
NM_001291701.1 | NP_001278630.1 | 7 | 1141 | |||
NM_012312.4 | NP_036444.1 | 8 | 1192 | |||
GenBank mRNA | AY366243.1 | - | 8 | 1167 | ||
L41347.1 | - | 8 | 1176 | |||
L76667.1 | - | 8 | 874 | |||
L76668.1 | - | 7 | 868 | |||
U24079.1 | - | 8 | 1155 |
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 8 | 1224 | ||
NM_001281972.1 | NP_001268901.1 | 7 | 1173 | |||
NM_012314.5 | NP_036446.3 | 9 | 1246 | |||
GenBank mRNA | AF002255.1 | - | 9 | 1203 | ||
AY388472.1 | - | 8 | 1195 | |||
BC144566.1 | - | 8 | 1246 | 9053094 | ||
BC171788.1 | - | 8 | 1246 | 9054442 | ||
U24077.1 | - | 8 | 1148 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 10 | 1478 | ||
NM_001282170.1 | NP_001269099.1 | 9 | 1193 | |||
NM_001282171.1 | NP_001269100.1 | 8 | 1157 | |||
GenBank mRNA | AK301140.1 | - | 10 | 1478 | ||
L76661.1 | - | 10 | 1451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
