siRNA Details
Gene Symbol: CDK11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313896.1 | NP_001300825.1 | 3 | 461 | ||
NM_001313982.1 | NP_001300911.1 | 3 | 461 | |||
NM_024011.3 | NP_076916.2 | 3 | 461 | |||
NM_033529.3 | NP_277071.2 | 3 | 461 | |||
NR_132739.1 | 3 | 461 | ||||
NR_132740.1 | 3 | 461 | ||||
GenBank mRNA | AF067518.1 | - | 3 | 219 | ||
AF067519.1 | - | 3 | 219 | |||
AF067520.1 | - | 3 | 219 | |||
AF067521.1 | - | 3 | 219 | |||
AF067522.1 | - | 3 | 219 | |||
AF067523.1 | - | 3 | 219 | |||
AF067526.1 | - | 3 | 237 | |||
AF067527.1 | - | 3 | 237 | |||
AF067528.1 | - | 3 | 386 | |||
AF067529.1 | - | 3 | 386 | |||
AK301495.1 | - | 3 | 223 | |||
AK303446.1 | - | 3 | 219 | |||
AK303451.1 | - | 3 | 232 | |||
BC028187.1 | - | 3 | 340 | |||
DC388505.1 | - | 3 | 248 | |||
U07704.1 | - | 3 | 251 | |||
U07705.1 | - | 3 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
