siRNA Details
Gene Symbol: USP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017414.3 | NP_059110.2 | 3, 4 | 588 | ||
XM_006724074.2 | XP_006724137.1 | 3, 4 | 659 | |||
GenBank mRNA | AF176642.2 | - | 2, 3 | 270 | ||
AJ243526.1 | - | 3, 4 | 453 | |||
AK313385.1 | - | 3, 4 | 409 | |||
AL136690.1 | - | 3, 4 | 409 | |||
BC014896.1 | - | 3, 4 | 394 | 3910720 | ||
BI752849.1 | - | 3, 4 | 484 | |||
BT006835.1 | - | 2, 3 | 250 | |||
CR457216.1 | - | 2, 3 | 250 | |||
CT841507.1 | - | 2, 3 | 316 |
Gene Symbol: USP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029164.1 | XP_016884653.1 | 3, 4 | 184 | ||
GenBank mRNA | AJ583822.1 | - | 2, 3 | 157 | ||
AJ586979.1 | - | 1, 2 | 94 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
