siRNA Details
Gene Symbol: GGTLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282879.1 | NP_001269808.1 | 2 | 82 | ||
NM_199127.2 | NP_954578.2 | 1 | 54 | |||
XM_005261815.3 | XP_005261872.2 | 2 | 111 | |||
XM_005261817.2 | XP_005261874.1 | 2 | 86 | |||
XM_005261818.2 | XP_005261875.1 | 2 | 111 | |||
XM_011530497.1 | XP_011528799.1 | 2 | 194 | |||
GenBank mRNA | BC069534.1 | - | 1 | 20 | 7262125 | |
BX282400.1 | - | 2 | 84 | |||
EL736203.1 | - | 1 | 20 | |||
HY050283.1 | - | 2 | 73 |
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288833.1 | NP_001275762.1 | 12 | 1851 | ||
NM_013421.2 | NP_038265.2 | 13 | 1779 | |||
NM_013430.2 | NP_038347.2 | 12 | 1539 | |||
GenBank mRNA | AL832738.1 | - | 1755 | |||
BC025927.1 | - | 10 | 1419 | 4548861 | ||
BC065921.1 | - | 2 | 62 | |||
BC069326.1 | - | 1 | 20 | |||
BC069473.1 | - | 1 | 20 | |||
BC069504.1 | - | 1 | 20 | |||
BC128234.1 | - | 1 | 48 | |||
BC128238.1 | - | 1 | 49 | |||
BC128239.1 | - | 1 | 49 | |||
CR456494.1 | - | 9 | 1129 | |||
DQ000492.2 | - | 3 | 234 | |||
J04131.1 | - | 13 | 1721 | |||
J05235.1 | - | 11 | 1818 | |||
L20490.1 | - | 2 | 59 | |||
L20493.1 | - | 1 | 208 | |||
M24087.1 | - | 8 | 1054 | |||
M24903.1 | - | 12 | 1539 | |||
X60069.1 | - | 10 | 1410 |
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261890.4 | XP_005261947.1 | 5 | 605 | ||
XM_006724392.3 | XP_006724455.1 | 9 | 1313 | |||
XM_006724395.3 | XP_006724458.1 | 6 | 765 | |||
XM_006724396.2 | XP_006724459.1 | 2 | 143 | |||
XM_011530621.2 | XP_011528923.1 | 2 | 130 | |||
XM_017029179.1 | XP_016884668.1 | 10 | 1994 | |||
XM_017029180.1 | XP_016884669.1 | 9 | 1452 | |||
XM_017029181.1 | XP_016884670.1 | 9 | 1890 | |||
XM_017029182.1 | XP_016884671.1 | 10 | 1994 | |||
XM_017029183.1 | XP_016884672.1 | 5 | 581 | |||
XM_017029184.1 | XP_016884673.1 | 2 | 153 | |||
XM_017029185.1 | XP_016884674.1 | 11 | 2856 | |||
XM_017029186.1 | XP_016884675.1 | 14 | 2457 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 9 | 1246 | |||
GenBank mRNA | BC143607.1 | - | 6 | 760 |
Gene Symbol: LOC102724823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030162.1 | XP_016885651.1 | 10 | 1790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
