siRNA Details
Gene Symbol: LOC102724197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724921.3 | XP_006724984.1 | 9 | 1822 | ||
XM_006724924.3 | XP_006724987.1 | 8 | 1664 | |||
XM_006724928.3 | XP_006724991.1 | 4 | 860 | |||
XM_006724929.2 | XP_006724992.1 | 2 | 209 | |||
XM_011546256.2 | XP_011544558.1 | 10 | 1956 | |||
XM_017030133.1 | XP_016885622.1 | 10 | 1387 | |||
XM_017030134.1 | XP_016885623.1 | 10 | 1390 | |||
XM_017030135.1 | XP_016885624.1 | 10 | 1815 | |||
XM_017030136.1 | XP_016885625.1 | 11 | 1857 | |||
XM_017030137.1 | XP_016885626.1 | 11 | 1854 | |||
XM_017030138.1 | XP_016885627.1 | 11 | 1377 | |||
XM_017030139.1 | XP_016885628.1 | 11 | 1380 | |||
XM_017030140.1 | XP_016885629.1 | 12 | 1847 | |||
XM_017030141.1 | XP_016885630.1 | 12 | 1844 | |||
XM_017030142.1 | XP_016885631.1 | 10 | 1688 | |||
XM_017030143.1 | XP_016885632.1 | 9 | 1823 | |||
XM_017030144.1 | XP_016885633.1 | 5 | 751 | |||
XM_017030145.1 | XP_016885634.1 | 6 | 1384 |
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288833.1 | NP_001275762.1 | 12 | 1917 | ||
NM_013421.2 | NP_038265.2 | 13 | 1845 | |||
NM_013430.2 | NP_038347.2 | 12 | 1605 | |||
GenBank mRNA | AL832738.1 | - | 1821 | |||
BC025927.1 | - | 10 | 1485 | 4548861 | ||
BC065921.1 | - | 2 | 128 | |||
BC069326.1 | - | 1 | 86 | |||
BC069473.1 | - | 1 | 86 | |||
BC069504.1 | - | 1 | 86 | |||
BC128234.1 | - | 1 | 114 | |||
BC128238.1 | - | 1 | 115 | |||
BC128239.1 | - | 1 | 115 | |||
CR456494.1 | - | 9 | 1195 | |||
DQ000492.2 | - | 3 | 300 | |||
J04131.1 | - | 13 | 1787 | |||
J05235.1 | - | 11 | 1884 | |||
L20490.1 | - | 2 | 125 | |||
M24087.1 | - | 8 | 1120 | |||
M24903.1 | - | 12 | 1605 | |||
X60069.1 | - | 10 | 1476 |
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261890.4 | XP_005261947.1 | 5 | 671 | ||
XM_006724392.3 | XP_006724455.1 | 9 | 1379 | |||
XM_006724395.3 | XP_006724458.1 | 6 | 831 | |||
XM_006724396.2 | XP_006724459.1 | 2 | 209 | |||
XM_011530621.2 | XP_011528923.1 | 2 | 196 | |||
XM_017029179.1 | XP_016884668.1 | 10 | 2060 | |||
XM_017029180.1 | XP_016884669.1 | 9 | 1518 | |||
XM_017029181.1 | XP_016884670.1 | 9 | 1956 | |||
XM_017029182.1 | XP_016884671.1 | 10 | 2060 | |||
XM_017029183.1 | XP_016884672.1 | 5 | 647 | |||
XM_017029184.1 | XP_016884673.1 | 2 | 219 | |||
XM_017029185.1 | XP_016884674.1 | 11 | 2922 | |||
XM_017029186.1 | XP_016884675.1 | 14 | 2523 | |||
GenBank mRNA | M30474.1 | - | 1 | 35 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 9 | 1312 |
Gene Symbol: GGTLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282879.1 | NP_001269808.1 | 2 | 148 | ||
NM_199127.2 | NP_954578.2 | 1 | 120 | |||
XM_005261815.3 | XP_005261872.2 | 2 | 177 | |||
XM_005261817.2 | XP_005261874.1 | 2 | 152 | |||
XM_005261818.2 | XP_005261875.1 | 2 | 177 | |||
XM_011530497.1 | XP_011528799.1 | 2 | 260 | |||
GenBank mRNA | BC069534.1 | - | 1 | 86 | 7262125 | |
BX282400.1 | - | 2 | 150 | |||
EL736203.1 | - | 1 | 86 | |||
HY050283.1 | - | 2 | 139 | |||
X98922.1 | - | 1 | 121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
