siRNA Details
Gene Symbol: LOC102724197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724921.3 | XP_006724984.1 | 12 | 2600 | ||
XM_006724924.3 | XP_006724987.1 | 11 | 2442 | |||
XM_006724928.3 | XP_006724991.1 | 7 | 1638 | |||
XM_006724929.2 | XP_006724992.1 | 5 | 987 | |||
XM_011546256.2 | XP_011544558.1 | 14 | 2653 | |||
XM_017030133.1 | XP_016885622.1 | 13 | 2165 | |||
XM_017030134.1 | XP_016885623.1 | 13 | 2168 | |||
XM_017030135.1 | XP_016885624.1 | 13 | 2593 | |||
XM_017030136.1 | XP_016885625.1 | 14 | 2635 | |||
XM_017030137.1 | XP_016885626.1 | 14 | 2632 | |||
XM_017030138.1 | XP_016885627.1 | 14 | 2155 | |||
XM_017030139.1 | XP_016885628.1 | 14 | 2158 | |||
XM_017030140.1 | XP_016885629.1 | 15 | 2625 | |||
XM_017030141.1 | XP_016885630.1 | 15 | 2622 | |||
XM_017030142.1 | XP_016885631.1 | 13 | 2466 | |||
XM_017030143.1 | XP_016885632.1 | 13 | 2520 | |||
XM_017030144.1 | XP_016885633.1 | 8 | 1529 | |||
XM_017030145.1 | XP_016885634.1 | 9 | 2162 |
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261890.4 | XP_005261947.1 | 8 | 1449 | ||
XM_006724392.3 | XP_006724455.1 | 12 | 2157 | |||
XM_006724395.3 | XP_006724458.1 | 9 | 1609 | |||
XM_006724396.2 | XP_006724459.1 | 5 | 987 | |||
XM_017029179.1 | XP_016884668.1 | 13 | 2838 | |||
XM_017029180.1 | XP_016884669.1 | 12 | 2296 | |||
XM_017029181.1 | XP_016884670.1 | 12 | 2734 | |||
XM_017029183.1 | XP_016884672.1 | 8 | 1425 | |||
XM_017029184.1 | XP_016884673.1 | 5 | 997 | |||
XM_017029186.1 | XP_016884675.1 | 18 | 3220 | |||
GenBank mRNA | M30474.1 | - | 5 | 702 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 13 | 2008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
