siRNA Details
Gene Symbol: INPP5F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014937.3 | NP_055752.1 | 8 | 1228 | ||
XM_006717720.3 | XP_006717783.1 | 8 | 3615 | |||
XM_011539524.2 | XP_011537826.1 | 8 | 991 | |||
XM_011539525.2 | XP_011537827.1 | 9 | 3698 | |||
XM_011539527.2 | XP_011537829.2 | 7 | 993 | |||
XM_011539528.2 | XP_011537830.1 | 7 | 1115 | |||
XM_017015935.1 | XP_016871424.1 | 8 | 982 | |||
XM_017015936.1 | XP_016871425.1 | 11 | 1616 | |||
GenBank mRNA | AB023183.1 | - | 8 | 1178 | ||
AK307758.1 | - | 8 | 1219 | |||
AL137528.1 | - | 2 | 193 | |||
BC067820.1 | - | 8 | 1189 | |||
BC111493.1 | - | 8 | 1139 | 6166990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
