siRNA Details
Gene Symbol: INPP5F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014937.3 | NP_055752.1 | 14, 15 | 1902 | ||
XM_006717720.3 | XP_006717783.1 | 14, 15 | 4289 | |||
XM_011539524.2 | XP_011537826.1 | 14, 15 | 1665 | |||
XM_011539525.2 | XP_011537827.1 | 15, 16 | 4372 | |||
XM_011539527.2 | XP_011537829.2 | 13, 14 | 1667 | |||
XM_011539528.2 | XP_011537830.1 | 13, 14 | 1789 | |||
XM_011539529.1 | XP_011537831.1 | 3, 4 | 323 | |||
XM_017015935.1 | XP_016871424.1 | 14, 15 | 1656 | |||
XM_017015936.1 | XP_016871425.1 | 17, 18 | 2290 | |||
GenBank mRNA | AB023183.1 | - | 14, 15 | 1852 | ||
AL137528.1 | - | 8, 9 | 867 | |||
BC111493.1 | - | 14, 15 | 1813 | 6166990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
