siRNA Details
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320893.1 | NP_001307822.1 | 3 | 586 | ||
NM_022552.4 | NP_072046.2 | 8 | 1272 | |||
NM_153759.3 | NP_715640.2 | 4 | 556 | |||
NM_175629.2 | NP_783328.1 | 8 | 1343 | |||
NR_135490.1 | 8 | 1343 | ||||
XM_005264175.4 | XP_005264232.1 | 8 | 1206 | |||
XM_005264177.4 | XP_005264234.1 | 3 | 498 | |||
XM_011532662.2 | XP_011530964.1 | 6 | 1208 | |||
XM_011532663.2 | XP_011530965.1 | 9 | 1385 | |||
XM_011532664.2 | XP_011530966.1 | 8 | 1283 | |||
XM_011532666.2 | XP_011530968.1 | 4 | 571 | |||
XM_011532667.2 | XP_011530969.1 | 3 | 388 | |||
XM_017003526.1 | XP_016859015.1 | 8 | 1314 | |||
XM_017003527.1 | XP_016859016.1 | 3 | 542 | |||
GenBank mRNA | AB208833.1 | - | 8 | 1242 | ||
AF067972.2 | - | 8 | 1234 | |||
AF331856.1 | - | 8 | 1222 | |||
AF480163.1 | - | 4 | 552 | |||
BC043617.1 | - | 8 | 1242 | 6150112 | ||
BM470515.1 | - | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
