siRNA Details
Gene Symbol: PRR5-ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181334.5 | NP_851851.3 | 5 | 656 | ||
GenBank mRNA | AF195968.1 | - | 4 | 247 | ||
AF195969.1 | - | 3 | 162 | |||
AL355192.1 | - | 8 | 748 | |||
BC132755.1 | - | 3 | 147 | 40146777 | ||
BC132757.1 | - | 3 | 147 | 40146779 | ||
BC144382.1 | - | 3 | 147 | 9052903 | ||
BX335168.2 | - | 5 | 619 |
Gene Symbol: ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017526.1 | NP_001017526.1 | 2 | 127 | ||
NM_001198726.1 | NP_001185655.1 | 2 | 127 | |||
NM_181335.2 | NP_851852.2 | 2 | 127 | |||
GenBank mRNA | AK001306.1 | - | 2 | 127 | ||
AK022305.1 | - | 1 | 69 | |||
BC010490.2 | - | 1 | 59 | |||
BC048280.1 | - | 2 | 79 | 5190702 | ||
BC059382.1 | - | 2 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
