siRNA Details
Gene Symbol: HIBCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222979.1 | - | 3 | 157 | ||
AK223023.1 | - | 3 | 157 | |||
AK308042.1 | - | 3 | 157 | |||
BC005190.1 | - | 3 | 210 | |||
DB034438.1 | - | 3 | 383 | |||
U66669.1 | - | 3 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
