siRNA Details
Gene Symbol: HAUS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017518.7 | NP_059988.3 | 10 | 1222 | ||
NR_073156.1 | 13 | 1992 | ||||
GenBank mRNA | AF267739.1 | - | 10 | 1153 | ||
AK125219.1 | - | 13 | 1987 | |||
AL122036.1 | - | 5 | 1790 | |||
BC008141.1 | - | 10 | 1222 | 4154976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
