siRNA Details
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003294.3 | NP_003285.2 | 4 | 377 | ||
GenBank mRNA | AF206665.1 | - | 3 | 335 | ||
AF206666.1 | - | 2 | 253 | |||
AF206667.1 | - | 2 | 139 | |||
BC038416.1 | - | 3 | 331 | |||
BC051852.1 | - | 3 | 330 | |||
BC065923.1 | - | 3 | 536 | 5752632 | ||
BC074974.2 | - | 4 | 358 | 30915512 | ||
BC074975.2 | - | 4 | 358 | 30915294 | ||
FJ931118.1 | - | 2 | 174 | |||
M30038.1 | - | 3 | 342 | |||
M33491.1 | - | 3 | 321 | |||
M37488.1 | - | 3 | 330 |
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024164.5 | NP_077078.5 | 4 | 348 | ||
GenBank mRNA | AK308208.1 | - | 4 | 348 | ||
BC028059.1 | - | 3 | 337 | 5181815 | ||
BC029356.1 | - | 4 | 348 | 4695458 | ||
FJ931119.1 | - | 2 | 178 | |||
FJ931120.1 | - | 2 | 178 | |||
M33492.1 | - | 3 | 323 | |||
M33493.1 | - | 3 | 302 | |||
S55551.1 | - | 3 | 332 |
Gene Symbol: TPSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012217.2 | NP_036349.1 | 3 | 355 | ||
GenBank mRNA | AF206664.1 | - | 2 | 192 | ||
AF421357.1 | - | 2 | 202 | |||
AY055427.1 | - | 2 | 144 | |||
BC069143.1 | - | 3 | 346 | 7217003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
