siRNA Details
Gene Symbol: RSC1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006511.2 | NP_006502.1 | 1 | 335 | ||
GenBank mRNA | CR999543.1 | - | 1 | 110 |
Gene Symbol: DDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032341.4 | NP_115717.3 | 10 | 1670 | ||
GenBank mRNA | AL598343.1 | - | 4 | 506 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023544, NM_001017966, AI854132, BC031491, BC150752, and others.
Gene Ontology Categories:
