siRNA Details
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278554.1 | NP_001265483.1 | 4 | 646 | ||
NM_001278555.1 | NP_001265484.1 | 4 | 454 | |||
NM_006357.3 | NP_006348.1 | 4 | 685 | |||
NM_182678.2 | NP_872619.1 | 5 | 486 | |||
XM_005246244.2 | XP_005246301.1 | 4 | 642 | |||
GenBank mRNA | AB017644.1 | - | 4 | 328 | ||
AF085362.1 | - | 4 | 386 | |||
AK314145.1 | - | 4 | 656 | |||
BC003554.1 | - | 4 | 368 | 3537280 | ||
BC092407.1 | - | 4 | 625 | 6180242 | ||
BI257596.1 | - | 4 | 450 | |||
BQ932163.1 | - | 5 | 486 | |||
BT019345.1 | - | 3 | 267 | |||
DA568152.1 | - | 4 | 580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
