siRNA Details
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007352.3 | NP_031378.1 | 4 | 400 | ||
GenBank mRNA | AK315798.1 | - | 4 | 269 | ||
BC005216.1 | - | 4 | 274 | 3949903 | ||
BI760781.1 | - | 4 | 399 | |||
BM129235.1 | - | 3 | 180 | |||
M16630.1 | - | 4 | 245 | |||
M18692.1 | - | 4 | 272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
