siRNA Details
Gene Symbol: RTEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283009.1 | NP_001269938.1 | 34 | 4375 | ||
NM_001283010.1 | NP_001269939.1 | 33 | 4096 | |||
NM_016434.3 | NP_057518.1 | 34 | 4375 | |||
NM_032957.4 | NP_116575.3 | 34 | 4447 | |||
GenBank mRNA | AF217795.1 | - | 34 | 4375 | ||
AK000485.1 | - | 10 | 1578 | |||
AK302508.1 | - | 34 | 3966 | |||
AK304798.1 | - | 33 | 3592 | |||
AL080127.1 | - | 11 | 1481 | |||
JX119186.1 | - | 33 | 3548 |
Gene Symbol: RTEL1-TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037882.1 | 34 | 4375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
