siRNA Details
Gene Symbol: SDHAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003264.2 | 13 | 1903 | |||
GenBank mRNA | AF088032.1 | - | 1 | 14 | ||
AK300421.1 | - | 9 | 1244 |
Gene Symbol: SDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294332.1 | NP_001281261.1 | 11 | 1597 | ||
NM_004168.3 | NP_004159.2 | 12 | 1741 | |||
XM_011514072.1 | XP_011512374.1 | 12 | 1741 | |||
XM_017009685.1 | XP_016865174.1 | 12 | 1741 | |||
GenBank mRNA | AB208991.1 | - | 12 | 1627 | ||
AK094879.1 | - | 7 | 1562 | |||
AK131350.1 | - | 3 | 2355 | |||
AK131478.1 | - | 2 | 3831 | |||
AK225027.1 | - | 12 | 1642 | |||
AK291311.1 | - | 12 | 1640 | |||
AK295937.1 | - | 11 | 1502 | |||
AK302524.1 | - | 12 | 1648 | |||
BC001380.2 | - | 12 | 1637 | 3051442 | ||
BC041016.1 | - | 10 | 1245 | 3138724 | ||
D30648.1 | - | 12 | 1614 | |||
DQ402982.1 | - | 10 | 1413 | |||
L21936.1 | - | 12 | 1632 |
Gene Symbol: SDHAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003265.3 | 12 | 1671 | |||
GenBank mRNA | AK057553.1 | - | 11 | 1501 | ||
AK074764.1 | - | 12 | 1617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
