siRNA Details
Gene Symbol: NDUFS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184979.1 | NP_001171908.1 | 2 | 309 | ||
NM_004552.2 | NP_004543.1 | 2 | 313 | |||
GenBank mRNA | AF020352.1 | - | 2 | 263 | ||
AF047434.1 | - | 2 | 274 | |||
AF086268.1 | - | 2 | 264 | |||
AK312188.1 | - | 2 | 287 | |||
BC001884.1 | - | 2 | 231 | 3544747 | ||
BC093057.1 | - | 2 | 253 | 30382835 | ||
CR456904.1 | - | 1 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
