siRNA Details
Gene Symbol: KIR3DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242867.1 | NP_001229796.1 | 4 | 656 | ||
NM_006737.3 | NP_006728.2 | 4 | 656 | |||
XM_011526940.2 | XP_011525242.1 | 4 | 780 | |||
XM_017026784.1 | XP_016882273.1 | 4 | 780 | |||
GenBank mRNA | AF208683.1 | - | 2 | 560 | ||
AF208684.1 | - | 2 | 560 | |||
AF208685.1 | - | 2 | 560 | |||
AF208686.1 | - | 2 | 560 | |||
AF208687.1 | - | 2 | 560 | |||
AF262965.1 | - | 2 | 542 | |||
AF262966.1 | - | 2 | 542 | |||
AF262967.1 | - | 4 | 656 | |||
AF263617.1 | - | 4 | 656 | |||
AJ276125.1 | - | 4 | 624 | |||
AW951147.1 | - | 1 | 281 | |||
AY059418.1 | - | 2 | 554 | |||
AY059419.1 | - | 2 | 554 | |||
AY059420.1 | - | 2 | 554 | |||
AY366256.2 | - | 4 | 635 | |||
AY789067.1 | - | 4 | 623 | |||
AY789068.1 | - | 4 | 623 | |||
AY789069.1 | - | 3 | 593 | |||
AY789070.1 | - | 4 | 623 | |||
AY789071.1 | - | 3 | 593 | |||
BC105678.1 | - | 4 | 648 | 40004817 | ||
BC107951.1 | - | 4 | 648 | 40004820 | ||
DQ256504.1 | - | 2 | 555 | |||
EF582384.1 | - | 4 | 623 | |||
L41270.1 | - | 4 | 642 | |||
L76665.1 | - | 4 | 634 | |||
L76666.1 | - | 4 | 634 | |||
U30272.1 | - | 4 | 625 | |||
X93595.1 | - | 4 | 653 | |||
X93596.1 | - | 4 | 623 | |||
X94373.1 | - | 4 | 624 | |||
X94374.1 | - | 4 | 623 |
Gene Symbol: 1060P11.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ630586.1 | - | 4 | 624 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 3 | 367 | ||
GenBank mRNA | AF285434.1 | - | 2 | 298 | ||
AY366242.1 | - | 3 | 367 | |||
KP260924.1 | - | 3 | 334 | |||
L76669.1 | - | 3 | 349 | |||
U24075.1 | - | 3 | 352 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 3 | 375 | ||
GenBank mRNA | AF022048.1 | - | 3 | 338 | ||
AF135556.1 | - | 2 | 275 | |||
AF135566.1 | - | 1 | 275 | |||
AF285435.1 | - | 2 | 298 | |||
AY789057.1 | - | 3 | 338 | |||
BC032422.1 | - | 3 | 351 | 5220339 | ||
BC050730.1 | - | 3 | 369 | 5219336 | ||
BC110613.1 | - | 3 | 352 | 40031619 | ||
FN395076.1 | - | 3 | 338 | |||
FN552550.1 | - | 3 | 320 | |||
L41268.1 | - | 3 | 375 | |||
L76662.1 | - | 3 | 349 | |||
L76663.1 | - | 3 | 349 | |||
U24074.1 | - | 3 | 359 | |||
U73395.1 | - | 3 | 359 | |||
U96190.1 | - | 1 | 275 |
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 3 | 351 | ||
GenBank mRNA | AF022046.1 | - | 3 | 351 | ||
AM999888.1 | - | 3 | 346 | |||
X89892.1 | - | 3 | 358 | |||
X98858.1 | - | 3 | 338 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 3 | 373 | ||
NM_001291701.1 | NP_001278630.1 | 3 | 373 | |||
NM_012312.4 | NP_036444.1 | 3 | 373 | |||
GenBank mRNA | AF285438.1 | - | 2 | 298 | ||
AJ002102.1 | - | 3 | 338 | |||
AJ002103.1 | - | 3 | 338 | |||
AJ002104.1 | - | 3 | 338 | |||
AJ002105.1 | - | 3 | 338 | |||
AM409094.1 | - | 3 | 339 | |||
AY366243.1 | - | 3 | 348 | |||
AY366244.1 | - | 3 | 337 | |||
BC108917.1 | - | 3 | 340 | 40017299 | ||
BC119631.1 | - | 3 | 357 | 40115568 | ||
BC119632.1 | - | 3 | 357 | 40115570 | ||
BC125146.1 | - | 2 | 322 | |||
BC126856.1 | - | 3 | 340 | 40017301 | ||
FR750994.1 | - | 3 | 373 | |||
L41347.1 | - | 3 | 357 | |||
U24079.1 | - | 3 | 336 | |||
X89893.1 | - | 3 | 338 |
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 3 | 427 | ||
NM_001281972.1 | NP_001268901.1 | 3 | 427 | |||
NM_012314.5 | NP_036446.3 | 3 | 427 | |||
GenBank mRNA | AF002255.1 | - | 3 | 384 | ||
AF135564.1 | - | 2 | 179 | |||
AJ417554.1 | - | 2 | 318 | |||
AJ417555.1 | - | 2 | 318 | |||
AJ620615.2 | - | 2 | 311 | |||
AJ620616.2 | - | 2 | 311 | |||
AY102623.1 | - | 3 | 338 | |||
AY102624.1 | - | 1 | 50 | |||
AY366245.1 | - | 3 | 372 | |||
AY366246.1 | - | 3 | 345 | |||
AY388472.1 | - | 3 | 376 | |||
AY789064.2 | - | 3 | 322 | |||
BC096694.1 | - | 3 | 349 | 40026979 | ||
BC101977.1 | - | 3 | 349 | 40004312 | ||
BC103693.1 | - | 3 | 349 | 40004314 | ||
BC108918.1 | - | 3 | 340 | 40017300 | ||
BC125147.1 | - | 2 | 322 | |||
BC127084.1 | - | 3 | 355 | 40120468 | ||
BC144566.1 | - | 3 | 427 | 9053094 | ||
BC171788.1 | - | 3 | 427 | 9054442 | ||
L76671.1 | - | 3 | 349 | |||
U24077.1 | - | 3 | 329 | |||
U96188.1 | - | 1 | 275 | |||
X94609.1 | - | 3 | 393 |
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF272389.1 | - | 3 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
