siRNA Details
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002000.3 | NP_001991.1 | 2 | 256 | ||
NM_133269.3 | NP_579803.1 | 2 | 256 | |||
NM_133271.3 | NP_579805.1 | 2 | 256 | |||
XM_011526625.2 | XP_011524927.1 | 2 | 180 | |||
XM_017026473.1 | XP_016881962.1 | 1 | 25 | |||
GenBank mRNA | AK290922.1 | - | 2 | 114 | ||
BC027953.1 | - | 2 | 83 | 5211453 | ||
CV029531.1 | - | 2 | 59 | |||
D87853.1 | - | 2 | 59 | |||
D87854.1 | - | 2 | 59 | |||
D87858.1 | - | 2 | 59 | |||
D87859.1 | - | 2 | 59 | |||
DQ075334.1 | - | 2 | 59 | |||
DQ075335.1 | - | 2 | 59 | |||
DQ075336.1 | - | 2 | 59 | |||
DQ075337.1 | - | 2 | 59 | |||
DQ075338.1 | - | 2 | 59 | |||
DQ075339.1 | - | 2 | 59 | |||
S82919.1 | - | 2 | 59 | |||
U43677.1 | - | 2 | 64 | |||
U43774.1 | - | 2 | 64 | |||
U56236.1 | - | 2 | 69 | |||
X92560.1 | - | 2 | 59 | |||
X92561.1 | - | 2 | 59 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
