siRNA Details
Gene Symbol: DYNLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281729.1 | NP_001268658.1 | 2 | 90 | ||
NM_001319157.1 | NP_001306086.1 | 2 | 90 | |||
NM_014183.3 | NP_054902.1 | 2 | 90 | |||
NM_177953.2 | NP_808852.1 | 3 | 169 | |||
NR_104032.1 | 2 | 90 | ||||
GenBank mRNA | AF132750.1 | - | 2 | 70 | ||
AF161511.1 | - | 2 | 61 | |||
AF165516.1 | - | 2 | 43 | |||
AF178431.1 | - | 1 | 15 | |||
AK026864.1 | - | 1 | 26 | |||
AK294214.1 | - | 2 | 48 | |||
AY026513.1 | - | 1 | 15 | |||
BC002481.1 | - | 2 | 68 | 3347555 | ||
BC007223.2 | - | 1 | 12 | |||
BG110161.1 | - | 2 | 75 | |||
BG419592.1 | - | 2 | 59 | |||
BM561321.1 | - | 2 | 60 | |||
BM791453.1 | - | 3 | 151 | |||
BU163605.1 | - | 109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
