siRNA Details
Gene Symbol: CDK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000075.3 | NP_000066.1 | 4 | 679 | ||
GenBank mRNA | AK297901.1 | - | 3 | 422 | ||
AK310724.1 | - | 3 | 753 | |||
AK313701.1 | - | 4 | 503 | |||
BC003644.2 | - | 4 | 526 | 3532812 | ||
BC005864.2 | - | 4 | 590 | 2961345 | ||
BC010153.2 | - | 4 | 528 | 3531300 | ||
BI832658.1 | - | 4 | 679 | |||
CR407668.1 | - | 3 | 387 | |||
CR542247.1 | - | 3 | 387 | |||
M14505.1 | - | 4 | 614 | |||
U79269.1 | - | 4 | 474 | |||
Z48970.1 | - | 4 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
