siRNA Details
Gene Symbol: BIRC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012271.1 | NP_001012271.1 | 4 | 471 | ||
NM_001168.2 | NP_001159.2 | 3 | 402 | |||
GenBank mRNA | AB028869.1 | - | 4 | 376 | ||
AB154416.1 | - | 3 | 281 | |||
AF077350.1 | - | 3 | 305 | |||
AK223428.1 | - | 3 | 347 | |||
AK301793.1 | - | 3 | 355 | |||
AK311917.1 | - | 3 | 306 | |||
AY830084.1 | - | 281 | ||||
BC008718.2 | - | 3 | 314 | 2961114 | ||
BC034148.1 | - | 3 | 325 | 4656567 | ||
BC065497.1 | - | 3 | 295 | 5394399 | ||
BE537971.1 | - | 4 | 382 | |||
BI520186.1 | - | 4 | 481 | |||
BQ687404.1 | - | 3 | 179 | |||
CB112371.1 | - | 4 | 404 | |||
CR541740.1 | - | 3 | 281 | |||
DQ508245.1 | - | 4 | 286 | |||
DQ508246.1 | - | 4 | 282 | |||
DQ508248.1 | - | 4 | 314 | |||
DQ508249.1 | - | 3 | 931 | |||
HM625836.1 | - | 3 | 321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
